The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020632.2 | NP_065683.2 | 7 - 8 | 794 | 83 |
NM_130840.2 | NP_570855.2 | 6 - 7 | 691 | 83 | |
NM_130841.2 | NP_570856.2 | 6 - 7 | 749 | 83 | |
XM_005250393.1 | XP_005250450.1 | 8 - 9 | 1141 | 83 | |
XM_005250394.3 | XP_005250451.1 | 6 - 7 | 1053 | 83 | |
GenBank mRNA | AF245517.1 | - | 7 - 8 | 779 | 83 |
AK055789.1 | - | 2 - 3 | 182 | 83 | |
AK292945.1 | - | 7 - 8 | 794 | 83 | |
BC109304.1 | - | 6 - 7 | 599 | 83 | |
BC109305.1 | - | 6 - 7 | 599 | 83 |