The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257189.1 | NP_001244118.1 | 1 - 2 | 704 | 60 |
NM_019082.3 | NP_061955.1 | 1 - 2 | 704 | 60 | |
GenBank mRNA | AF247666.1 | - | 1 - 2 | 94 | 60 |
AJ131712.1 | - | 1 - 2 | 128 | 60 | |
AK022774.1 | - | 1 - 2 | 75 | 60 | |
AK125595.1 | - | 1 - 2 | 1219 | 60 | |
AK222662.1 | - | 1 - 2 | 93 | 60 | |
AK309276.1 | - | 1 - 2 | 74 | 60 | |
AK315363.1 | - | 1 - 2 | 93 | 60 | |
AL136700.1 | - | 1 - 2 | 70 | 60 | |
BC001235.1 | - | 1 - 2 | 91 | 60 | |
CR457213.1 | - | 1 - 2 | 61 | 60 |