The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019082.3 | NP_061955.1 | 7 - 8 | 1651 | 115 |
GenBank mRNA | AF247666.1 | - | 7 - 8 | 1041 | 115 |
AJ131712.1 | - | 7 - 8 | 1075 | 115 | |
AK022774.1 | - | 7 - 8 | 1022 | 115 | |
AK125595.1 | - | 7 - 8 | 2166 | 115 | |
AK222662.1 | - | 7 - 8 | 1040 | 115 | |
AK309276.1 | - | 6 - 7 | 860 | 115 | |
AL136700.1 | - | 7 - 8 | 1017 | 115 | |
BC001235.1 | - | 7 - 8 | 1038 | 115 | |
CR457213.1 | - | 7 - 8 | 1008 | 115 |