The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024838.4 | NP_079114.3 | 3 - 3 | 326 | 120 |
XM_005252597.2 | XP_005252654.1 | 3 - 3 | 402 | 120 | |
XM_017016665.1 | XP_016872154.1 | 4 - 4 | 412 | 120 | |
GenBank mRNA | AK125249.1 | - | 3 - 3 | 254 | 120 |
BC035132.2 | - | 3 - 3 | 247 | 120 | |
BX647989.1 | - | 3 - 3 | 239 | 120 | |
DA496769.1 | - | 3 - 3 | 339 | 120 |