Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CR619427.1 |
- |
1 |
1426 |
|
Chr. 10 - 3783669 - 3785196 [-] on Build GRCh38 |
94 |
GenBankmRNA |
BX395555.2 |
- |
1 |
|
|
Chr. 10 - 3783694 - 3784740 [+] on Build GRCh38 |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105376360 |
LOC105376360 |
|
-32.9kb
|
Anti-sense |
Chr. - 3318695 - 3750764 on Build GRCh38 |
Downstream |
uncharacterized LOC105376365 |
LOC105376365 |
|
48.8kb
|
Anti-sense |
Chr. - 3833950 - 3834728 on Build GRCh38 |
View Full Product Details