The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303457.1 | NP_001290386.1 | 2 - 3 | 2413 | 65 |
NM_014657.2 | NP_055472.1 | 3 - 4 | 2553 | 65 | |
XM_011529114.1 | XP_011527416.1 | 2 - 3 | 2413 | 65 | |
XM_017028148.1 | XP_016883637.1 | 2 - 3 | 2413 | 65 | |
GenBank mRNA | AB007866.2 | - | 2 - 3 | 2502 | 65 |
AK308164.1 | - | 2 - 3 | 2402 | 65 | |
BC013121.1 | - | 3 - 4 | 2516 | 65 | |
BC013755.1 | - | 2 - 3 | 2366 | 65 | |
BC107778.1 | - | 2 - 3 | 2410 | 65 | |
BC131759.1 | - | 1 - 2 | 88 | 65 |