The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015638.2 | NP_056453.1 | 11 - 12 | 1442 | 68 |
NM_199368.1 | NP_955400.1 | 11 - 12 | 1418 | 68 | |
XM_011528772.2 | XP_011527074.1 | 11 - 12 | 1464 | 68 | |
XM_011528774.1 | XP_011527076.1 | 6 - 7 | 755 | 68 | |
XM_017027799.1 | XP_016883288.1 | 11 - 12 | 1464 | 68 | |
XM_017027800.1 | XP_016883289.1 | 10 - 11 | 1256 | 68 | |
XM_017027801.1 | XP_016883290.1 | 9 - 10 | 920 | 68 | |
GenBank mRNA | AF055022.1 | - | 2 - 3 | 145 | 68 |
AK074106.1 | - | 11 - 12 | 1421 | 68 | |
AK298958.1 | - | 10 - 11 | 1389 | 68 | |
AK301368.1 | - | 3 - 4 | 361 | 68 | |
AK303476.1 | - | 10 - 11 | 1317 | 68 | |
AK315582.1 | - | 11 - 12 | 1434 | 68 | |
AL117480.1 | - | 5 - 6 | 698 | 68 | |
BC013144.1 | - | 11 - 12 | 1411 | 68 |