The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015638.2 | NP_056453.1 | 12 - 13 | 1543 | 66 |
NM_199368.1 | NP_955400.1 | 12 - 13 | 1519 | 66 | |
XM_011528774.1 | XP_011527076.1 | 7 - 8 | 856 | 66 | |
XM_017027799.1 | XP_016883288.1 | 12 - 13 | 1565 | 66 | |
XM_017027800.1 | XP_016883289.1 | 11 - 12 | 1357 | 66 | |
XM_017027801.1 | XP_016883290.1 | 10 - 11 | 1021 | 66 | |
GenBank mRNA | AF055022.1 | - | 3 - 4 | 246 | 66 |
AK074106.1 | - | 12 - 13 | 1522 | 66 | |
AK298958.1 | - | 11 - 12 | 1490 | 66 | |
AK301368.1 | - | 4 - 5 | 462 | 66 | |
AK303476.1 | - | 11 - 12 | 1418 | 66 | |
AK315582.1 | - | 12 - 13 | 1535 | 66 | |
AL117480.1 | - | 6 - 7 | 799 | 66 | |
BC001323.1 | - | 1 - 2 | 56 | 66 | |
BC013144.1 | - | 12 - 13 | 1512 | 66 |