The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015638.2 | NP_056453.1 | 9 - 10 | 1252 | 69 |
NM_199368.1 | NP_955400.1 | 9 - 10 | 1228 | 69 | |
XM_011528772.2 | XP_011527074.1 | 9 - 10 | 1274 | 69 | |
XM_017027799.1 | XP_016883288.1 | 9 - 10 | 1274 | 69 | |
XM_017027800.1 | XP_016883289.1 | 8 - 9 | 1066 | 69 | |
GenBank mRNA | AK074106.1 | - | 9 - 10 | 1231 | 69 |
AK298958.1 | - | 8 - 9 | 1199 | 69 | |
AK303476.1 | - | 8 - 9 | 1127 | 69 | |
AK315582.1 | - | 9 - 10 | 1244 | 69 | |
AL117480.1 | - | 3 - 4 | 508 | 69 | |
BC013144.1 | - | 9 - 10 | 1221 | 69 |