Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TRPC4AP
- Entrez Gene ID
-
26133
- Gene Name
- transient receptor potential cation channel subfamily C member 4 associated protein
- Gene Aliases
-
C20orf188, PPP1R158, TRRP4AP, TRUSS
- Chromosome Location
-
Chr.20: 35002404 - 35092837 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK074106.1 |
- |
16 - 17 |
3599 |
91 |
View Full Product Details