Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OVOL2
Entrez Gene ID
58495
Gene Name
ovo like zinc finger 2
Gene Aliases
EUROIMAGE566589, PPCD1, ZNF339
Chromosome Location
Chr.20: 18024152 - 18059188 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021220.3 NP_067043.2 1 - 2 346 151
GenBank mRNA AK022284.1 - 1 - 2 341 151
BC006148.1 - 1 - 2 346 151
BT007295.1 - 1 - 2 98 151

View Full Product Details