The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172129.1 | NP_001165600.1 | 12 - 13 | 1619 | 57 |
NM_001172130.1 | NP_001165601.1 | 12 - 13 | 1616 | 57 | |
NM_001172131.1 | NP_001165602.1 | 12 - 13 | 1616 | 57 | |
NM_001172132.1 | NP_001165603.1 | 13 - 14 | 1685 | 57 | |
NM_001172133.1 | NP_001165604.1 | 13 - 14 | 1683 | 57 | |
NM_002110.3 | NP_002101.2 | 12 - 13 | 1619 | 57 | |
GenBank mRNA | AK026432.1 | - | 12 - 13 | 1553 | 57 |
AK225819.1 | - | 12 - 13 | 1565 | 57 | |
AK289896.1 | - | 12 - 13 | 1607 | 57 | |
AK290928.1 | - | 13 - 14 | 1631 | 57 | |
AK298726.1 | - | 13 - 14 | 1629 | 57 | |
AK314335.1 | - | 12 - 13 | 1532 | 57 | |
BC014435.1 | - | 12 - 13 | 1505 | 57 | |
BC094847.1 | - | 12 - 13 | 1516 | 57 | |
BC108930.1 | - | 12 - 13 | 1546 | 57 | |
BC108931.1 | - | 12 - 13 | 1546 | 57 | |
BC113854.1 | - | 12 - 13 | 1543 | 57 | |
BC114463.1 | - | 12 - 13 | 1546 | 57 | |
M16592.1 | - | 11 - 12 | 1394 | 57 |