Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HCK
Entrez Gene ID
3055
Gene Name
HCK proto-oncogene, Src family tyrosine kinase
Gene Aliases
JTK9, p59Hck, p61Hck
Chromosome Location
Chr.20: 32052188 - 32101856 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172129.1 NP_001165600.1 5 - 6 664 62
NM_001172130.1 NP_001165601.1 5 - 6 661 62
NM_001172131.1 NP_001165602.1 5 - 6 661 62
NM_001172132.1 NP_001165603.1 6 - 7 730 62
NM_001172133.1 NP_001165604.1 6 - 7 728 62
NM_002110.3 NP_002101.2 5 - 6 664 62
GenBank mRNA AK225819.1 - 5 - 6 610 62
AK289896.1 - 5 - 6 652 62
AK290928.1 - 6 - 7 676 62
AK298726.1 - 6 - 7 674 62
AK314335.1 - 5 - 6 577 62
BC014435.1 - 5 - 6 550 62
BC094847.1 - 5 - 6 561 62
BC108930.1 - 5 - 6 591 62
BC108931.1 - 5 - 6 591 62
BC113854.1 - 5 - 6 588 62
BC114463.1 - 5 - 6 591 62
M16592.1 - 4 - 5 439 62

View Full Product Details