The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003091.3 | NP_003082.1 | 1 - 2 | 172 | 102 |
NM_198216.1 | NP_937859.1 | 1 - 2 | 172 | 102 | |
GenBank mRNA | AK301034.1 | - | 1 - 2 | 104 | 102 |
AK301206.1 | - | 1 - 2 | 106 | 102 | |
AK307093.1 | - | 1 - 2 | 106 | 102 | |
AK307266.1 | - | 1 - 2 | 106 | 102 | |
AK312620.1 | - | 1 - 2 | 107 | 102 | |
BC080516.1 | - | 1 - 2 | 90 | 102 | |
BM559678.1 | - | - | 172 | 102 | |
X17567.1 | - | 1 - 2 | 63 | 102 | |
X17568.1 | - | 1 - 2 | 48 | 102 |