Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SNRPB
- Entrez Gene ID
-
6628
- Gene Name
- small nuclear ribonucleoprotein polypeptides B and B1
- Gene Aliases
-
CCMS, COD, SNRPB1, Sm-B/B', SmB/B', SmB/SmB', snRNP-B
- Chromosome Location
-
Chr.20: 2461635 - 2470853 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK301034.1 |
- |
3 - 4 |
378 |
79 |
View Full Product Details