The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173512.1 | NP_001166983.1 | 1 - 2 | 339 | 70 |
NM_138777.3 | NP_620132.1 | 1 - 2 | 339 | 70 | |
XM_011519186.1 | XP_011517488.1 | 1 - 2 | 87 | 70 | |
XM_017015287.1 | XP_016870776.1 | 1 - 2 | 87 | 70 | |
GenBank mRNA | AK291809.1 | - | 1 - 2 | 68 | 70 |
AK292790.1 | - | 1 - 2 | 68 | 70 | |
AK298046.1 | - | 1 - 2 | 58 | 70 | |
AK300718.1 | - | 1 - 2 | 42 | 70 | |
BF982227.1 | - | - | 74 | 70 |