The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272003.1 | NP_001258932.1 | 26 - 27 | 3096 | 60 |
NM_020822.2 | NP_065873.2 | 27 - 28 | 3231 | 60 | |
XM_011518877.2 | XP_011517179.1 | 27 - 28 | 7201 | 60 | |
XM_011518878.2 | XP_011517180.1 | 27 - 28 | 7210 | 60 | |
XM_011518879.2 | XP_011517181.1 | 27 - 28 | 7201 | 60 | |
XM_011518880.1 | XP_011517182.1 | 26 - 27 | 3058 | 60 | |
XM_011518881.2 | XP_011517183.1 | 24 - 25 | 4118 | 60 | |
XM_017014931.1 | XP_016870420.1 | 25 - 26 | 3106 | 60 | |
XM_017014932.1 | XP_016870421.1 | 26 - 27 | 2998 | 60 | |
XM_017014933.1 | XP_016870422.1 | 24 - 25 | 2825 | 60 | |
GenBank mRNA | AB037843.1 | - | 27 - 28 | 3224 | 60 |
AK127272.1 | - | 26 - 27 | 3096 | 60 | |
BC136618.1 | - | 27 - 28 | 3203 | 60 | |
BC171770.1 | - | 27 - 28 | 3203 | 60 |