Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GOLGA2
Entrez Gene ID
2801
Gene Name
golgin A2
Gene Aliases
GM130
Chromosome Location
Chr.9: 128255829 - 128276007 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004486.4 NP_004477.3 23 - 24 2547 93
XM_005251930.1 XP_005251987.1 24 - 25 2646 93
XM_005251931.1 XP_005251988.1 23 - 24 2565 93
XM_005251932.1 XP_005251989.1 22 - 23 2477 93
GenBank mRNA AF248953.1 - 23 - 24 2498 93
AY823636.1 - 1 - 2 110 93
BC014188.2 - 11 - 12 1564 93
BC069268.1 - 23 - 24 2547 93
BX647920.1 - 5 - 6 93
L06147.1 - 9 - 10 1406 93

View Full Product Details