The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004486.4 | NP_004477.3 | 7 - 8 | 573 | 147 |
XM_005251930.1 | XP_005251987.1 | 8 - 9 | 672 | 147 | |
GenBank mRNA | AF248953.1 | - | 7 - 8 | 524 | 147 |
BC069268.1 | - | 7 - 8 | 573 | 147 |