The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318878.1 | NP_001305807.1 | 4 - 5 | 824 | 69 |
NM_001318879.1 | NP_001305808.1 | 4 - 5 | 685 | 69 | |
NM_173575.3 | NP_775846.2 | 4 - 5 | 786 | 69 | |
NR_134911.1 | 4 - 5 | 463 | 69 | ||
XM_011539688.1 | XP_011537990.1 | 4 - 5 | 857 | 69 | |
XM_011539690.2 | XP_011537992.1 | 4 - 5 | 772 | 69 | |
XM_011539693.2 | XP_011537995.1 | 4 - 5 | 790 | 69 | |
XM_011539694.1 | XP_011537996.1 | 4 - 5 | 524 | 69 | |
XM_011539695.1 | XP_011537997.1 | 4 - 5 | 725 | 69 | |
XM_011539696.1 | XP_011537998.1 | 4 - 5 | 532 | 69 | |
GenBank mRNA | AK057849.1 | - | 4 - 5 | 463 | 69 |
AK098366.1 | - | 3 - 4 | 623 | 69 | |
AK302102.1 | - | 4 - 5 | 771 | 69 | |
AK308956.1 | - | 4 - 5 | 863 | 69 | |
AK310725.1 | - | 4 - 5 | 790 | 69 | |
AY098866.1 | - | 4 - 5 | 786 | 69 | |
BC045760.1 | - | 4 - 5 | 675 | 69 |