The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318878.1 | NP_001305807.1 | 6 - 7 | 955 | 97 |
NM_001318879.1 | NP_001305808.1 | 6 - 7 | 816 | 97 | |
NM_173575.3 | NP_775846.2 | 6 - 7 | 917 | 97 | |
NR_134911.1 | 6 - 7 | 594 | 97 | ||
XM_011539688.1 | XP_011537990.1 | 6 - 7 | 988 | 97 | |
XM_011539690.2 | XP_011537992.1 | 6 - 7 | 903 | 97 | |
XM_011539693.2 | XP_011537995.1 | 6 - 7 | 921 | 97 | |
XM_011539694.1 | XP_011537996.1 | 6 - 7 | 655 | 97 | |
XM_011539695.1 | XP_011537997.1 | 6 - 7 | 856 | 97 | |
XM_011539696.1 | XP_011537998.1 | 6 - 7 | 663 | 97 | |
GenBank mRNA | AK057849.1 | - | 6 - 7 | 594 | 97 |
AK302102.1 | - | 6 - 7 | 902 | 97 | |
AK308956.1 | - | 6 - 7 | 994 | 97 | |
AY098866.1 | - | 6 - 7 | 917 | 97 | |
BC045760.1 | - | 6 - 7 | 806 | 97 |