Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP63
Entrez Gene ID
80254
Gene Name
centrosomal protein 63
Gene Aliases
SCKL6
Chromosome Location
Chr.3: 134485733 - 134629607 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042400.1 NP_001035859.1 9 - 10 1243 63
NM_025180.3 NP_079456.2 10 - 11 1519 63
XM_005247795.4 XP_005247852.1 9 - 10 1192 63
XM_005247797.3 XP_005247854.1 9 - 10 1447 63
XM_005247799.3 XP_005247856.1 9 - 10 1260 63
XM_006713760.3 XP_006713823.1 9 - 10 1124 63
XM_011513194.2 XP_011511496.1 9 - 10 1241 63
XM_011513197.2 XP_011511499.1 8 - 9 1075 63
XM_017007244.1 XP_016862733.1 9 - 10 1235 63
XM_017007250.1 XP_016862739.1 9 - 10 1233 63
XM_017007251.1 XP_016862740.1 9 - 10 1231 63
XM_017007252.1 XP_016862741.1 9 - 10 1166 63
XM_017007253.1 XP_016862742.1 9 - 10 1227 63
XM_017007254.1 XP_016862743.1 9 - 10 1124 63
XM_017007255.1 XP_016862744.1 9 - 10 1447 63
XM_017007256.1 XP_016862745.1 10 - 11 1497 63
XM_017007257.1 XP_016862746.1 9 - 10 1229 63
XM_017007258.1 XP_016862747.1 9 - 10 1124 63
XM_017007259.1 XP_016862748.1 9 - 10 1241 63
GenBank mRNA AK023448.1 - 9 - 10 1189 63
AK056465.1 - 10 - 11 1497 63
BC068997.1 - 9 - 10 1163 63

View Full Product Details