The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004815.3 | NP_004806.3 | 13 - 14 | 1801 | 116 |
XM_006711048.3 | XP_006711111.1 | 12 - 13 | 1435 | 116 | |
XM_011542438.2 | XP_011540740.1 | 13 - 14 | 1676 | 116 | |
XM_011542439.2 | XP_011540741.1 | 13 - 14 | 1521 | 116 | |
XM_011542440.2 | XP_011540742.1 | 12 - 13 | 1395 | 116 | |
GenBank mRNA | AB208873.1 | - | 12 - 13 | 1407 | 116 |
AK307733.1 | - | 13 - 14 | 1630 | 116 | |
AK310491.1 | - | 13 - 14 | 1521 | 116 | |
AL833445.1 | - | 1 - 2 | 6010 | 116 | |
BC093741.1 | - | 12 - 13 | 1474 | 116 | |
BC093767.1 | - | 12 - 13 | 1477 | 116 | |
U90920.1 | - | 13 - 14 | 1615 | 116 |