Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUCKS1
Entrez Gene ID
64710
Gene Name
nuclear casein kinase and cyclin dependent kinase substrate 1
Gene Aliases
JC7, NUCKS
Chromosome Location
Chr.1: 205712819 - 205750244 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022731.4 NP_073568.2 2 - 3 343 90
XM_005245453.1 XP_005245510.1 2 - 3 319 90
GenBank mRNA AB049824.1 - 2 - 3 274 90
AJ012584.1 - 2 - 3 170 90
AK025133.1 - 2 - 3 138 90
AK225228.1 - 2 - 3 138 90
AK300416.1 - 2 - 3 277 90
BC000805.1 - 2 - 3 264 90
BQ638451.1 - 2 - 3 375 90
CR457339.1 - 2 - 3 72 90
DB004374.1 - 2 - 3 333 90

View Full Product Details