Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUCKS1
Entrez Gene ID
64710
Gene Name
nuclear casein kinase and cyclin dependent kinase substrate 1
Gene Aliases
JC7, NUCKS
Chromosome Location
Chr.1: 205712819 - 205750244 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022731.4 NP_073568.2 3 - 4 446 90
XM_005245453.1 XP_005245510.1 3 - 4 422 90
GenBank mRNA AB049824.1 - 3 - 4 377 90
AJ012584.1 - 3 - 4 273 90
AK025133.1 - 3 - 4 241 90
AK225228.1 - 3 - 4 241 90
AK300416.1 - 3 - 4 380 90
BC000805.1 - 3 - 4 367 90
BQ638451.1 - 3 - 4 478 90
CR457339.1 - 3 - 4 175 90
DB004374.1 - 3 - 4 436 90

View Full Product Details