The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308190.1 | NP_001295119.1 | 3 - 4 | 321 | 68 |
NM_002099.7 | NP_002090.4 | 4 - 5 | 420 | 68 | |
XM_017008134.1 | XP_016863623.1 | 4 - 5 | 424 | 68 | |
XM_017008135.1 | XP_016863624.1 | 5 - 6 | 487 | 68 | |
XM_017008136.1 | XP_016863625.1 | 3 - 4 | 389 | 68 | |
GenBank mRNA | AK290561.1 | - | 4 - 5 | 385 | 68 |
AY950614.1 | - | 4 - 5 | 269 | 68 | |
BC005319.1 | - | 4 - 5 | 326 | 68 | |
BC013328.1 | - | 4 - 5 | 307 | 68 | |
DA537533.1 | - | 4 - 5 | 385 | 68 | |
DA539682.1 | - | 4 - 5 | 421 | 68 | |
EU338231.1 | - | 5 - 6 | 389 | 68 | |
EU338232.1 | - | 4 - 5 | 306 | 68 | |
EU338233.1 | - | 5 - 6 | 389 | 68 | |
EU338234.1 | - | 3 - 4 | 213 | 68 | |
GU347002.1 | - | 5 - 6 | 413 | 68 | |
GU347003.1 | - | 5 - 6 | 492 | 68 | |
GU347004.1 | - | 5 - 6 | 470 | 68 | |
GU347006.1 | - | 4 - 5 | 312 | 68 | |
GU347008.1 | - | 3 - 4 | 207 | 68 | |
L07251.1 | - | 3 - 4 | 173 | 68 | |
M36281.1 | - | 4 - 5 | 279 | 68 | |
U00177.1 | - | 4 - 5 | 269 | 68 | |
U00178.1 | - | 3 - 4 | 173 | 68 | |
U00179.1 | - | 2 - 3 | 74 | 68 | |
X08054.1 | - | 4 - 5 | 325 | 68 | |
X51798.1 | - | 4 - 5 | 293 | 68 |