The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006346.2 | NP_006337.2 | 1 - 2 | 255 | 145 |
XM_011534881.2 | XP_011533183.1 | 1 - 2 | 283 | 145 | |
XM_011534882.2 | XP_011533184.1 | 1 - 2 | 283 | 145 | |
XM_011534884.2 | XP_011533186.1 | 1 - 2 | 283 | 145 | |
XM_011534886.2 | XP_011533188.1 | 1 - 2 | 283 | 145 | |
XM_017020351.1 | XP_016875840.1 | 1 - 2 | 283 | 145 | |
XM_017020352.1 | XP_016875841.1 | 1 - 2 | 209 | 145 | |
XM_017020353.1 | XP_016875842.1 | 1 - 2 | 206 | 145 | |
XM_017020354.1 | XP_016875843.1 | 1 - 2 | 206 | 145 | |
GenBank mRNA | AK309828.1 | - | 1 - 2 | 212 | 145 |
AY370776.1 | - | 1 - 2 | 154 | 145 | |
BC006009.1 | - | 1 - 2 | 208 | 145 | |
BC008432.1 | - | 1 - 2 | 208 | 145 | |
BC041806.1 | - | 1 - 2 | 207 | 145 | |
BC051911.1 | - | 1 - 2 | 185 | 145 | |
BX341357.2 | - | 1 - 2 | 255 | 145 |