The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020846.1 | NP_065897.1 | 6 - 7 | 1503 | 101 |
GenBank mRNA | AB040891.1 | - | 6 - 7 | 1473 | 101 |
AK026476.1 | - | 6 - 7 | 1050 | 101 | |
AK291006.1 | - | 6 - 7 | 1633 | 101 | |
BC031691.2 | - | 4 - 5 | 823 | 101 | |
BC040993.1 | - | 6 - 7 | 1782 | 101 |