Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHROOM3
Entrez Gene ID
57619
Gene Name
shroom family member 3
Gene Aliases
APXL3, MSTP013, SHRM, ShrmL
Chromosome Location
Chr.4: 76435100 - 76783253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020859.3 NP_065910.3 5 - 6 4702 70
XM_005263163.4 XP_005263220.1 4 - 5 9156 70
XM_005263164.4 XP_005263221.1 4 - 5 3714 70
XM_005263165.4 XP_005263222.1 4 - 5 5140 70
XM_006714282.3 XP_006714345.1 5 - 6 3857 70
XM_011532157.2 XP_011530459.2 4 - 5 4544 70
XM_011532158.2 XP_011530460.1 6 - 7 4503 70
XM_011532159.1 XP_011530461.1 5 - 6 4692 70
XM_011532160.1 XP_011530462.1 3 - 4 3704 70
XM_011532161.2 XP_011530463.1 4 - 5 3882 70
XM_017008488.1 XP_016863977.1 5 - 6 3860 70
XM_017008489.1 XP_016863978.1 5 - 6 3761 70
GenBank mRNA AB040914.1 - 1 - 2 1901 70
AB055660.1 - 5 - 6 4703 70
AK128677.1 - 1 - 2 3204 70
BC007291.1 - 1 - 2 63 70

View Full Product Details