Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSD17B13
Entrez Gene ID
345275
Gene Name
hydroxysteroid 17-beta dehydrogenase 13
Gene Aliases
HMFN0376, NIIL497, SCDR9, SDR16C3
Chromosome Location
Chr.4: 87303789 - 87322906 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178135.4 NP_835236.2 2 - 3 388 85
GenBank mRNA AB073347.1 - 2 - 3 357 85
AK292784.1 - 2 - 3 362 85
AY186249.1 - 2 - 3 368 85
AY268355.1 - 2 - 3 338 85
AY358575.1 - 2 - 3 388 85
BC112303.1 - 2 - 3 347 85
BC112305.1 - 2 - 3 347 85
DR004209.1 - 2 - 3 393 85

View Full Product Details