The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136230.2 | NP_001129702.1 | 3 - 4 | 517 | 75 |
NM_178135.4 | NP_835236.2 | 4 - 5 | 625 | 75 | |
GenBank mRNA | AB073347.1 | - | 4 - 5 | 594 | 75 |
AK292784.1 | - | 4 - 5 | 599 | 75 | |
AY186249.1 | - | 4 - 5 | 605 | 75 | |
AY186250.1 | - | 3 - 4 | 497 | 75 | |
AY268355.1 | - | 4 - 5 | 575 | 75 | |
AY358575.1 | - | 4 - 5 | 625 | 75 | |
BC112303.1 | - | 4 - 5 | 584 | 75 | |
BC112305.1 | - | 4 - 5 | 584 | 75 | |
DR004209.1 | - | 4 - 5 | 630 | 75 |