The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286734.1 | NP_001273663.1 | 2 - 3 | 238 | 157 |
NM_001813.2 | NP_001804.2 | 2 - 3 | 238 | 157 | |
XM_011531544.2 | XP_011529846.1 | 2 - 3 | 231 | 157 | |
XM_011531545.2 | XP_011529847.1 | 2 - 3 | 237 | 157 | |
XM_011531546.2 | XP_011529848.1 | 2 - 3 | 237 | 157 | |
XM_011531547.2 | XP_011529849.1 | 2 - 3 | 237 | 157 | |
XM_011531548.2 | XP_011529850.1 | 2 - 3 | 237 | 157 | |
XM_011531549.2 | XP_011529851.1 | 2 - 3 | 237 | 157 | |
XM_017007659.1 | XP_016863148.1 | 2 - 3 | 237 | 157 | |
GenBank mRNA | AB209996.1 | - | 2 - 3 | 163 | 157 |
AU100312.1 | - | 2 - 3 | 179 | 157 | |
Z15005.1 | - | 2 - 3 | 238 | 157 |