Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CENPE
Entrez Gene ID
1062
Gene Name
centromere protein E
Gene Aliases
CENP-E, KIF10, MCPH13, PPP1R61
Chromosome Location
Chr.4: 103105806 - 103198409 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286734.1 NP_001273663.1 39 - 40 6444 131
NM_001813.2 NP_001804.2 41 - 42 6807 131
XM_011531544.2 XP_011529846.1 40 - 41 6725 131
XM_011531545.2 XP_011529847.1 40 - 41 6647 131
XM_011531546.2 XP_011529848.1 40 - 41 6602 131
XM_011531547.2 XP_011529849.1 40 - 41 6518 131
XM_011531548.2 XP_011529850.1 40 - 41 6515 131
XM_011531549.2 XP_011529851.1 39 - 40 6362 131
XM_017007659.1 XP_016863148.1 39 - 40 6356 131
GenBank mRNA AA210728.1 - 2 - 3 131
AB209996.1 - 39 - 40 6369 131
Z15005.1 - 42 - 43 6693 131

View Full Product Details