The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286734.1 | NP_001273663.1 | 46 - 47 | 7737 | 79 |
NM_001813.2 | NP_001804.2 | 48 - 49 | 8100 | 79 | |
XM_011531544.2 | XP_011529846.1 | 47 - 48 | 8018 | 79 | |
XM_011531545.2 | XP_011529847.1 | 47 - 48 | 7940 | 79 | |
XM_011531546.2 | XP_011529848.1 | 47 - 48 | 7895 | 79 | |
XM_011531547.2 | XP_011529849.1 | 47 - 48 | 7811 | 79 | |
XM_011531548.2 | XP_011529850.1 | 47 - 48 | 7808 | 79 | |
XM_011531549.2 | XP_011529851.1 | 46 - 47 | 7655 | 79 | |
XM_017007659.1 | XP_016863148.1 | 46 - 47 | 7649 | 79 | |
GenBank mRNA | AB209996.1 | - | 46 - 47 | 7662 | 79 |
BQ447425.1 | - | 1 - 2 | 79 | ||
Z15005.1 | - | 49 - 50 | 7986 | 79 |