The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193314.1 | NP_001180243.1 | 1 - 2 | 548 | 69 |
NM_022067.3 | NP_071350.2 | 2 - 3 | 419 | 69 | |
XM_017021579.1 | XP_016877068.1 | 1 - 2 | 273 | 69 | |
XM_017021581.1 | XP_016877070.1 | 1 - 2 | 273 | 69 | |
GenBank mRNA | AK098254.1 | - | 1 - 2 | 509 | 69 |
BC015054.1 | - | 2 - 3 | 340 | 69 |