The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007214.4 | NP_009145.1 | 8 - 9 | 1003 | 143 |
XM_011535399.2 | XP_011533701.1 | 8 - 9 | 752 | 143 | |
XM_017010218.1 | XP_016865707.1 | 8 - 9 | 836 | 143 | |
GenBank mRNA | AF100141.1 | - | 8 - 9 | 734 | 143 |
AJ011779.1 | - | 8 - 9 | 832 | 143 | |
AK001465.1 | - | 9 - 10 | 866 | 143 | |
AK027374.1 | - | 5 - 6 | 374 | 143 | |
AK074835.1 | - | 8 - 9 | 819 | 143 | |
AL137338.1 | - | 5 - 6 | 350 | 143 | |
BC023598.2 | - | 8 - 9 | 914 | 143 | |
BC047221.1 | - | 8 - 9 | 872 | 143 | |
BC048287.1 | - | 8 - 9 | 859 | 143 | |
KU178551.1 | - | 8 - 9 | 734 | 143 |