The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001858.5 | NP_001849.2 | 40 - 41 | 2687 | 62 |
XM_011535437.2 | XP_011533739.1 | 40 - 41 | 2954 | 62 | |
XM_011535438.2 | XP_011533740.1 | 40 - 41 | 2682 | 62 | |
XM_017010253.1 | XP_016865742.1 | 39 - 40 | 2912 | 62 | |
XM_017010254.1 | XP_016865743.1 | 39 - 40 | 2900 | 62 | |
XM_017010255.1 | XP_016865744.1 | 40 - 41 | 2893 | 62 | |
XM_017010256.1 | XP_016865745.1 | 39 - 40 | 2645 | 62 | |
XM_017010257.1 | XP_016865746.1 | 39 - 40 | 2828 | 62 | |
XM_017010258.1 | XP_016865747.1 | 39 - 40 | 2561 | 62 | |
XM_017010259.1 | XP_016865748.1 | 40 - 41 | 2953 | 62 | |
GenBank mRNA | BC113362.1 | - | 40 - 41 | 2655 | 62 |
BC113364.1 | - | 40 - 41 | 2655 | 62 | |
D38163.1 | - | 40 - 41 | 2687 | 62 | |
U09279.1 | - | 8 - 9 | 359 | 62 |