The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040455.1 | NP_001035545.1 | 7 - 8 | 846 | 60 |
GenBank mRNA | AF151799.1 | - | 7 - 8 | 888 | 60 |
AK292501.1 | - | 7 - 8 | 1326 | 60 | |
AK292974.1 | - | 7 - 8 | 837 | 60 | |
AK303023.1 | - | 7 - 8 | 802 | 60 | |
AY358442.1 | - | 7 - 8 | 855 | 60 | |
BC114522.1 | - | 7 - 8 | 875 | 60 |