Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MED14
Entrez Gene ID
9282
Gene Name
mediator complex subunit 14
Gene Aliases
CRSP150, CRSP2, CSRP, CXorf4, DRIP150, EXLM1, RGR1, TRAP170
Chromosome Location
Chr.X: 40649543 - 40736122 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004229.3 NP_004220.2 9 - 10 1315 86
XM_005272701.2 XP_005272758.1 9 - 10 1225 86
XM_011544000.2 XP_011542302.1 10 - 11 1197 86
XM_017029962.1 XP_016885451.1 10 - 11 1190 86
GenBank mRNA AB006651.1 - 9 - 10 1294 86
AF104256.1 - 9 - 10 1175 86
AF135802.1 - 9 - 10 1248 86
AF304448.1 - 9 - 10 1294 86
AK293070.1 - 9 - 10 1493 86
AK308871.1 - 9 - 10 1123 86
BC098377.1 - 9 - 10 1315 86
BC132672.1 - 9 - 10 1284 86
BC132674.1 - 9 - 10 1284 86

View Full Product Details