The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198474.3 | NP_940876.2 | 1 - 2 | 640 | 66 |
XM_005252878.4 | XP_005252935.1 | 2 - 3 | 417 | 66 | |
GenBank mRNA | AK298137.1 | - | 2 - 3 | 235 | 66 |
AY358591.1 | - | 1 - 2 | 242 | 66 | |
BC035895.1 | - | 1 - 2 | 467 | 66 | |
BC092488.1 | - | 1 - 2 | 485 | 66 | |
BX647908.1 | - | 1 - 2 | 528 | 66 | |
DB257515.1 | - | 1 - 2 | 508 | 66 |