Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LVRN
Entrez Gene ID
206338
Gene Name
laeverin
Gene Aliases
APQ, AQPEP, TAQPEP
Chromosome Location
Chr.5: 115962440 - 116027602 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK027809.1 - 1 - 2 168 147

View Full Product Details