The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033510.2 | NP_277045.1 | 5 - 6 | 809 | 62 |
XM_011522125.2 | XP_011520427.1 | 6 - 7 | 2980 | 62 | |
XM_017022687.1 | XP_016878176.1 | 6 - 7 | 2980 | 62 | |
GenBank mRNA | AB051529.2 | - | 5 - 6 | 814 | 62 |
BC151833.1 | - | 5 - 6 | 814 | 62 |