Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM227B
Entrez Gene ID
196951
Gene Name
family with sequence similarity 227 member B
Gene Aliases
C15orf33
Chromosome Location
Chr.15: 49326997 - 49620931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005254219.3 XP_005254276.1 12 - 13 1484 125
GenBank mRNA AK023699.1 - 1 - 2 87 125

View Full Product Details