Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRP1B
Entrez Gene ID
53353
Gene Name
LDL receptor related protein 1B
Gene Aliases
LRP-1B, LRP-DIT, LRPDIT
Chromosome Location
Chr.2: 140231423 - 142132463 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018557.2 NP_061027.2 85 - 86 14114 115
XM_017004341.1 XP_016859830.1 85 - 86 18690 115
XM_017004342.1 XP_016859831.1 56 - 57 8113 115
GenBank mRNA AB209707.1 - 32 - 33 4507 115
AF176832.1 - 85 - 86 14114 115

View Full Product Details