Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DMBT1
- Entrez Gene ID
-
1755
- Gene Name
- deleted in malignant brain tumors 1
- Gene Aliases
-
GP340, SAG, muclin
- Chromosome Location
-
Chr.10: 122560665 - 122643736 on Build GRCh38
View Full Product Details