The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039467.1 | NP_001034556.1 | 4 - 5 | 401 | 104 |
NM_005873.2 | NP_005864.1 | 4 - 5 | 501 | 104 | |
XM_005260182.3 | XP_005260239.1 | 3 - 4 | 749 | 104 | |
XM_005260183.2 | XP_005260240.1 | 3 - 4 | 336 | 104 | |
XM_011528484.1 | XP_011526786.1 | 3 - 4 | 1931 | 104 | |
XM_011528485.2 | XP_011526787.1 | 4 - 5 | 980 | 104 | |
XM_011528486.2 | XP_011526788.1 | 4 - 5 | 433 | 104 | |
GenBank mRNA | AF493939.1 | - | 3 - 4 | 234 | 104 |
AK290081.1 | - | 4 - 5 | 518 | 104 | |
AK298240.1 | - | - | 416 | 104 | |
AY585188.1 | - | 3 - 4 | 234 | 104 | |
BC001318.1 | - | 3 - 4 | 316 | 104 | |
BC054337.1 | - | 4 - 5 | 444 | 104 | |
BC063010.1 | - | 4 - 5 | 489 | 104 | |
BT009804.1 | - | 3 - 4 | 234 | 104 | |
X91809.1 | - | 4 - 5 | 522 | 104 |