The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_126522.1 | 1 - 1 | 676 | 74 | |
GenBank mRNA | BC014011.2 | - | 1 - 1 | 611 | 74 |
BC029796.1 | - | 1 - 1 | 659 | 74 |