The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001624.3 | NP_001615.2 | 7 - 8 | 4108 | 99 |
XM_005266839.3 | XP_005266896.1 | 9 - 10 | 4946 | 99 | |
XM_017010333.1 | XP_016865822.1 | 7 - 8 | 4333 | 99 | |
GenBank mRNA | U83115.1 | - | 7 - 8 | 3363 | 99 |