The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012484.2 | NP_036616.2 | 3 - 4 | 406 | 114 |
GenBank mRNA | AF032862.1 | - | 3 - 4 | 260 | 114 |