Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSD17B4
Entrez Gene ID
3295
Gene Name
hydroxysteroid 17-beta dehydrogenase 4
Gene Aliases
DBP, MFE-2, MPF-2, PRLTS1, SDR8C1
Chromosome Location
Chr.5: 119452443 - 119542335 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000414.3 NP_000405.1 22 - 23 2125 81
NM_001199291.2 NP_001186220.1 23 - 24 2324 81
NM_001199292.1 NP_001186221.1 21 - 22 2071 81
NM_001292027.1 NP_001278956.1 23 - 24 2190 81
NM_001292028.1 NP_001278957.1 22 - 23 2250 81
GenBank mRNA AK094049.1 - 22 - 23 1994 81
AK293254.1 - 22 - 23 1997 81
AK295440.1 - 17 - 18 1737 81
AK298075.1 - 23 - 24 2252 81
AK299685.1 - 23 - 24 2095 81
AK301212.1 - 21 - 22 2017 81
AK308968.1 - 18 - 19 1839 81
AK312449.1 - 22 - 23 2071 81
BC003098.1 - 22 - 23 2059 81
CX782401.1 - 2 - 3 81
KU178135.1 - 22 - 23 1992 81
X87176.1 - 22 - 23 2040 81

View Full Product Details